missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzoic acid, 99%, Thermo Scientific Chemicals

Catalog Number p-3778600
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
456-22-4
C7H5FO2
140.11
MFCD00002530
BBYDXOIZLAWGSL-UHFFFAOYSA-N
p-fluorobenzoic acid, benzoic acid, 4-fluoro, para-fluorobenzoic acid, 4-fluoro-benzoic acid, benzoic acid, p-fluoro, unii-v5roo2hou4, p-fluoro-benzoic acid, v5roo2hou4, 4-fluorobenzoatd, p-flurobenzoic acid
9973
CHEBI:20364
4-fluorobenzoic acid
C1=CC(=CC=C1C(=O)O)F
This item is not returnable. View return policy
456-22-4
C7H5FO2
140.11
MFCD00002530
BBYDXOIZLAWGSL-UHFFFAOYSA-N
p-fluorobenzoic acid, benzoic acid, 4-fluoro, para-fluorobenzoic acid, 4-fluoro-benzoic acid, benzoic acid, p-fluoro, unii-v5roo2hou4, p-fluoro-benzoic acid, v5roo2hou4, 4-fluorobenzoatd, p-flurobenzoic acid
9973
CHEBI:20364
4-fluorobenzoic acid
C1=CC(=CC=C1C(=O)O)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 184.0°C to 187.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula FC6H4CO2H
Quantity 25 g
Beilstein 09, 333
Merck Index 15, 4203
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in alcohol and ether
Formula Weight 140.11
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluorobenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluorobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'