missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog Number p-3778595
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
349-88-2
C6H4ClFO2S
194.60
MFCD00007438
BFXHJFKKRGVUMU-UHFFFAOYSA-N
4-fluorobenzene-1-sulfonyl chloride, p-fluorobenzenesulfonyl chloride, 4-fluorobenzenesulphonyl chloride, benzenesulfonyl chloride, 4-fluoro, 4-fluorophenylsulfonyl chloride, 4-fluorobenzenesulfonic acid chloride, 4-fluorobenzenesulfonylchloride, benzenesulfonyl chloride, p-fluoro, 4-fluorobenzensulfonyl chloride, 4-fluoro-benzenesulfonyl chloride
9588
4-fluorobenzenesulfonyl chloride
FC1=CC=C(C=C1)S(Cl)(=O)=O
This item is not returnable. View return policy
349-88-2
C6H4ClFO2S
194.60
MFCD00007438
BFXHJFKKRGVUMU-UHFFFAOYSA-N
4-fluorobenzene-1-sulfonyl chloride, p-fluorobenzenesulfonyl chloride, 4-fluorobenzenesulphonyl chloride, benzenesulfonyl chloride, 4-fluoro, 4-fluorophenylsulfonyl chloride, 4-fluorobenzenesulfonic acid chloride, 4-fluorobenzenesulfonylchloride, benzenesulfonyl chloride, p-fluoro, 4-fluorobenzensulfonyl chloride, 4-fluoro-benzenesulfonyl chloride
9588
4-fluorobenzenesulfonyl chloride
FC1=CC=C(C=C1)S(Cl)(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 29°C to 34°C
Color Brown to White
Boiling Point 95°C to 96°C (2.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Linear Formula FC6H4SO2Cl
Quantity 25 g
Beilstein 11, 53
Formula Weight 194.61
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 4-Fluorobenzenesulfonyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluorobenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'