missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzaldehyde, 98+%, Thermo Scientific Chemicals

Catalog Number p-134270
26.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
459-57-4
C7H5FO
124.11
MFCD00003378
UOQXIWFBQSVDPP-UHFFFAOYSA-N
p-fluorobenzaldehyde, benzaldehyde, 4-fluoro, 4-fluoro-benzaldehyde, para-fluorobenzaldehyde, p-fluorbenzaldehyde, fluorobenzaldehyde 4-, benzaldehyde, p-fluoro, pfad, 4-fluoro benzaldehyde, para fluoro benzaldehyde
68023
4-fluorobenzaldehyde
C1=CC(=CC=C1C=O)F
This item is not returnable. View return policy
459-57-4
C7H5FO
124.11
MFCD00003378
UOQXIWFBQSVDPP-UHFFFAOYSA-N
p-fluorobenzaldehyde, benzaldehyde, 4-fluoro, 4-fluoro-benzaldehyde, para-fluorobenzaldehyde, p-fluorbenzaldehyde, fluorobenzaldehyde 4-, benzaldehyde, p-fluoro, pfad, 4-fluoro benzaldehyde, para fluoro benzaldehyde
68023
4-fluorobenzaldehyde
C1=CC(=CC=C1C=O)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point -10.0°C
Color Colorless to Yellow
Density 1.1500g/mL
Boiling Point 181.0°C (758.0 mmHg)
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4CHO
Refractive Index 1.5195 to 1.5215
Quantity 50 mL
Beilstein 07, I, 132
Specific Gravity 1.15
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in ether,acetone,benzene and alcohol
Formula Weight 124.11
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 4-Fluorobenzaldehyde, 98+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Fluorobenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'