missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number p-7022549
Click to view available options
Quantity:
25 g
100 g
500 g
459-57-4
C7H5FO
124.114
MFCD00003378
UOQXIWFBQSVDPP-UHFFFAOYSA-N
p-fluorobenzaldehyde, benzaldehyde, 4-fluoro, 4-fluoro-benzaldehyde, para-fluorobenzaldehyde, p-fluorbenzaldehyde, fluorobenzaldehyde 4-, benzaldehyde, p-fluoro, pfad, 4-fluoro benzaldehyde, para fluoro benzaldehyde
68023
4-fluorobenzaldehyde
C1=CC(=CC=C1C=O)F
459-57-4
C7H5FO
124.114
MFCD00003378
UOQXIWFBQSVDPP-UHFFFAOYSA-N
p-fluorobenzaldehyde, benzaldehyde, 4-fluoro, 4-fluoro-benzaldehyde, para-fluorobenzaldehyde, p-fluorbenzaldehyde, fluorobenzaldehyde 4-, benzaldehyde, p-fluoro, pfad, 4-fluoro benzaldehyde, para fluoro benzaldehyde
68023
4-fluorobenzaldehyde
C1=CC(=CC=C1C=O)F

4-Fluorobenzaldehyde is used in the preparation of pyrazolopyridine derivative, which finds application as mitogen-activated protein kinase (MAPK) inhibitor. It also serves as a synthetic intermediate in the preparation of pharmaceutical compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorobenzaldehyde is used in the preparation of pyrazolopyridine derivative, which finds application as mitogen-activated protein kinase (MAPK) inhibitor. It also serves as a synthetic intermediate in the preparation of pharmaceutical compounds.

Solubility
Immiscible with water.

Notes
Air sensitive. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY
Melting Point -10°C
Density 1.175
Boiling Point 180°C to 181°C
Flash Point 56°C (133°F)
Refractive Index 1.521
Quantity 25 g
UN Number UN1989
Beilstein 385857
Sensitivity Air Sensitive
Solubility Information Immiscible with water.
Formula Weight 124.11
Percent Purity 98%
Chemical Name or Material 4-Fluorobenzaldehyde
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'