missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoroaniline, 99%, Thermo Scientific Chemicals

Catalog Number p-7022705
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
371-40-4
C6H6FN
111.12
MFCD00007829
KRZCOLNOCZKSDF-UHFFFAOYSA-N
p-fluoroaniline, benzenamine, 4-fluoro, 1-amino-4-fluorobenzene, 4-fluorobenzenamine, p-fluorophenylamine, 4-fluoronaniline, aniline, p-fluoro, aniline, 4-fluoro, 4-fluoranilin, para-fluoroaniline
9731
CHEBI:28546
4-fluoroaniline
NC1=CC=C(F)C=C1
This item is not returnable. View return policy
371-40-4
C6H6FN
111.12
MFCD00007829
KRZCOLNOCZKSDF-UHFFFAOYSA-N
p-fluoroaniline, benzenamine, 4-fluoro, 1-amino-4-fluorobenzene, 4-fluorobenzenamine, p-fluorophenylamine, 4-fluoronaniline, aniline, p-fluoro, aniline, 4-fluoro, 4-fluoranilin, para-fluoroaniline
9731
CHEBI:28546
4-fluoroaniline
NC1=CC=C(F)C=C1

It finds its application as an analytical reagent in the enzymic detection of glucose. It is used as an intermediate in the manufacture of pharmaceuticals and plant growth regulators. It is employed as an intermediate for the production of Flumazenil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as an analytical reagent in the enzymic detection of glucose. It is used as an intermediate in the manufacture of pharmaceuticals and plant growth regulators. It is employed as an intermediate for the production of Flumazenil.

Solubility
Slightly soluble in water. Soluble in chloroform and methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Melting Point -2°C
Density 1.158
Boiling Point 186°C to 187°C
Flash Point 73°C (163°F)
Odor Recognizable
Refractive Index 1.54
Quantity 25 g
UN Number UN2941
Beilstein 742030
Merck Index 14,4169
Solubility Information Slightly soluble in water. Soluble in chloroform and methanol.
Formula Weight 111.12
Percent Purity 99%
Chemical Name or Material 4-Fluoroaniline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'