missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoroaniline, 98%, Thermo Scientific Chemicals
16.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | -1.9°C |
Color | Brown-Red to Yellow |
Density | 1.1570g/mL |
Boiling Point | 187.0°C |
Flash Point | 73°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Glass bottle |
Show More |
Safety and Handling
Product Identifier
- 4-Fluoroaniline
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
- H373-May cause damage to organs.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'