missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluoroacetophenone, 99%, Thermo Scientific Chemicals

Catalog Number p-134219
15.60 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
403-42-9
C8H7FO
138.14
MFCD00000354
ZDPAWHACYDRYIW-UHFFFAOYSA-N
4'-fluoroacetophenone, 4-fluoroacetophenone, 1-4-fluorophenyl ethanone, p-fluoroacetophenone, 1-4-fluorophenyl ethan-1-one, ethanone, 1-4-fluorophenyl, acetophenone, 4'-fluoro, 4 fluoroacetophenone, unii-tvq090602v, 1-acetyl-4-fluorobenzene
9828
1-(4-fluorophenyl)ethanone
CC(=O)C1=CC=C(F)C=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
403-42-9
C8H7FO
138.14
MFCD00000354
ZDPAWHACYDRYIW-UHFFFAOYSA-N
4'-fluoroacetophenone, 4-fluoroacetophenone, 1-4-fluorophenyl ethanone, p-fluoroacetophenone, 1-4-fluorophenyl ethan-1-one, ethanone, 1-4-fluorophenyl, acetophenone, 4'-fluoro, 4 fluoroacetophenone, unii-tvq090602v, 1-acetyl-4-fluorobenzene
9828
1-(4-fluorophenyl)ethanone
CC(=O)C1=CC=C(F)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 4°C
Color Colorless to Yellow
Density 1.1300g/mL
Boiling Point 196°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4COCH3
Refractive Index 1.5090 to 1.512
Quantity 25 mL
Specific Gravity 1.13
Formula Weight 138.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4'-Fluoroacetophenone, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4'-Fluoroacetophenone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'