missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-L-phenylglycine, 98%, Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Fluoro-L-phenylglycine is an optically active amino acid derivative that is used as an intermediate for making drugs.
Solubility
Soluble in 1M HCl.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
4-Fluoro-L-phenylglycine is an optically active amino acid derivative that is used as an intermediate for making drugs.
Solubility
Soluble in 1M HCl.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Melting Point | >300°C |
Color | White |
Beilstein | 5850631 |
Solubility Information | Soluble in 1M HCl. |
IUPAC Name | (2S)-2-azaniumyl-2-(4-fluorophenyl)acetate |
Formula Weight | 169.15 |
Percent Purity | 98% |
Quantity | 25 g |
Physical Form | Solid |
Chemical Name or Material | 4-Fluoro-L-phenylglycine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'