missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Fluoro-beta-nitrostyrene, 98%

Brand:  Alfa Aesar™ H64613.14

Code : NEW

Additional Details : CAS Number : 706-08-1 Weight : 0.02500kg Transport : UN number : 2811 Chem class : 1 Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 15411899

  • £96.60 / 25g


Estimated Shipment date
from Supplier 21-02-2018
Add to basket

Description

Description

Applications
It is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store at 99-102°C. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

4-Fluoro-beta-nitrostyrene
167.14
UN2811
1-Fluoro-4-(2-nitrovinyl)benzene; 1-(4-Fluorophenyl)-2-nitroethylene
706-08-1
MFCD00024824
2208372

Specifications

4-Fluoro-beta-nitrostyrene
99° to 102°C
706-08-1
98%
MFCD00024824
2208372
Pale yellow
25g
Crystalline Solid
167.14
UN2811
1-Fluoro-4-(2-nitrovinyl)benzene; 1-(4-Fluorophenyl)-2-nitroethylene
Videos
SDS