missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-5-nitrobenzene-1,2-diamine, Technical Grade, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 1,2-benzenediamine,4-fluoro-5-nitro, 1,2-benzenediamine, 4-fluoro-5-nitro, acmc-20ag30, 4,5-diamine-2-fluoronitrobenzene, 5-fluoro-4-nitrobenzene-1,2-diamine, 4-fluoro-5-nitro-1,2-phenylenediamine |
Formula Weight | 171.13 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | 4-Fluoro-5-nitrobenzene-1, 2-diamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'