missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-phenoxybenzaldehyde, 97%, Thermo Scientific™

Catalog Number p-7053628
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
68359-57-9
C13H9FO2
216.21
MFCD01318148
JDICMOLUAHZVDS-UHFFFAOYSA-N
4-fluoro-3-phenoxy-benzaldehyde, benzaldehyde, 4-fluoro-3-phenoxy, m-phenoxy-p-fluorobenzaldehyde, 2-fluoro-5-formyldiphenyl ether, 4-fluoro-3-phenoxy benzaldehyde, pubchem14130, 3-phenoxy-4fluorobenzaldehyde, ksc497q9b, 3-phenoxy-4-fluorobenzaldehyde, 3-phenoxy-4-fluoro-benzaldehyde
110068
4-fluoro-3-phenoxybenzaldehyde
FC1=CC=C(C=O)C=C1OC1=CC=CC=C1
This item is not returnable. View return policy
68359-57-9
C13H9FO2
216.21
MFCD01318148
JDICMOLUAHZVDS-UHFFFAOYSA-N
4-fluoro-3-phenoxy-benzaldehyde, benzaldehyde, 4-fluoro-3-phenoxy, m-phenoxy-p-fluorobenzaldehyde, 2-fluoro-5-formyldiphenyl ether, 4-fluoro-3-phenoxy benzaldehyde, pubchem14130, 3-phenoxy-4fluorobenzaldehyde, ksc497q9b, 3-phenoxy-4-fluorobenzaldehyde, 3-phenoxy-4-fluoro-benzaldehyde
110068
4-fluoro-3-phenoxybenzaldehyde
FC1=CC=C(C=O)C=C1OC1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 1.209 g/mL
Flash Point >110°C (230°F)
Refractive Index 1.583
Quantity 5 g
UN Number UN3082
Formula Weight 216.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Fluoro-3-phenoxybenzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'