missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-nitrobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7081006
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
453-71-4
C7H4FNO4
185.11
MFCD00007058
BOJWTAQWPVBIPG-UHFFFAOYSA-N
4-fluoro-3-nitrobenzoic acid, 3-nitro-4-fluorobenzoic acid, p-fluoro-3-nitrobenzoic acid, 4-fluoro-3-nitrobenzoicacid, 5-carboxy-2-fluoronitrobenzene, benzoic acid, 4-fluoro-3-nitro, 4-fluoro-3-nitro-benzoic acid, pubchem1306, acmc-1aqvm, 3-nitro-4-flurobenzoic acid
67987
4-fluoro-3-nitrobenzoic acid
OC(=O)C1=CC=C(F)C(=C1)[N+]([O-])=O
This item is not returnable. View return policy
453-71-4
C7H4FNO4
185.11
MFCD00007058
BOJWTAQWPVBIPG-UHFFFAOYSA-N
4-fluoro-3-nitrobenzoic acid, 3-nitro-4-fluorobenzoic acid, p-fluoro-3-nitrobenzoic acid, 4-fluoro-3-nitrobenzoicacid, 5-carboxy-2-fluoronitrobenzene, benzoic acid, 4-fluoro-3-nitro, 4-fluoro-3-nitro-benzoic acid, pubchem1306, acmc-1aqvm, 3-nitro-4-flurobenzoic acid
67987
4-fluoro-3-nitrobenzoic acid
OC(=O)C1=CC=C(F)C(=C1)[N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 121°C to 126°C
Boiling Point 92°C (15 mmHg)
Quantity 5 g
Beilstein 2107562
Formula Weight 185.11
Percent Purity 98%
Chemical Name or Material 4-Fluoro-3-nitrobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'