missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-3-nitrobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Fluoro-3-nitrobenzeneboronic acid is generally used as a intermediate in pharmaceutical and chemical research.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
4-Fluoro-3-nitrobenzeneboronic acid is generally used as a intermediate in pharmaceutical and chemical research.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications
Melting Point | 64°C to 65°C |
Color | Yellow |
Quantity | 1 g |
Beilstein | 20289173 |
Solubility Information | Slightly soluble in water. |
InChI Key | MOKWUYKZXFBLKR-UHFFFAOYSA-N |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)[N+](=O)[O-])F |
IUPAC Name | 2-(3-fluoro-4-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Molecular Weight (g/mol) | 267.063 |
Formula Weight | 267.06 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'