Learn More
Thermo Scientific Maybridge 4-Fluoro-3-nitrobenzaldehyde, 10g, 97%
Brand: Thermo Scientific Maybridge SB02056EA
4-Fluoro-3-nitrobenzaldehyde, 10g, 97%
Code : 31
Additional Details : CAS Number : 42564-51-2 Weight : 0.06000kg
Brand: Thermo Scientific Maybridge SB02056EA
4-Fluoro-3-nitrobenzaldehyde, 10g, 97%
Code : 31
Additional Details : CAS Number : 42564-51-2 Weight : 0.06000kg