missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-nitroaniline, 99%, ACROS Organics™

Brand:  Acros Organics 168400250

Code : 29

Additional Details : CAS Number : 364-76-1 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10645992

  • £19.00 / 25g


Estimated Shipment
06-02-2018
Add to basket


Specifications

Chemical Identifiers

4-Fluoro-3-nitroaniline
364-76-1
100.0
99%
FC6H3(NO2)NH2
MFCD00007833
99%
98.5
≥98.5%
C6H5FN2O2
156.12

Specifications

4-Fluoro-3-nitroaniline
96° to 99°C
Authentic
364-76-1
100.0
Powder
99%
FC6H3(NO2)NH2
MFCD00007833
Ochre
25g
99%
98.5
Glass bottle
≥98.5%
C6H5FN2O2
156.12
12, 729
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Fluoro-3-nitroaniline

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS