missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluoro-3'-nitroacetophenone, 98%, Thermo Scientific™

Catalog Number p-2690663
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
400-93-1
C8H6FNO3
183.14
MFCD00115369
PTCNZDJJIOLIKQ-UHFFFAOYSA-N
4'-fluoro-3'-nitroacetophenone, 1-4-fluoro-3-nitrophenyl ethanone, 4-fluoro-3-nitroacetophenone, 1-4-fluoro-3-nitrophenyl ethan-1-one, 1-4-fluoro-3-nitro-phenyl ethanone, 4-fluoro-3'-nitroacetophenone, ethanone, 1-4-fluoro-3-nitrophenyl, 4-fluoro-3-nitroacetophneone, 3'-nitro-4'-fluoroacetophenone
2734612
1-(4-fluoro-3-nitrophenyl)ethanone
CC(=O)C1=CC(=C(C=C1)F)[N+](=O)[O-]
This item is not returnable. View return policy
400-93-1
C8H6FNO3
183.14
MFCD00115369
PTCNZDJJIOLIKQ-UHFFFAOYSA-N
4'-fluoro-3'-nitroacetophenone, 1-4-fluoro-3-nitrophenyl ethanone, 4-fluoro-3-nitroacetophenone, 1-4-fluoro-3-nitrophenyl ethan-1-one, 1-4-fluoro-3-nitro-phenyl ethanone, 4-fluoro-3'-nitroacetophenone, ethanone, 1-4-fluoro-3-nitrophenyl, 4-fluoro-3-nitroacetophneone, 3'-nitro-4'-fluoroacetophenone
2734612
1-(4-fluoro-3-nitrophenyl)ethanone
CC(=O)C1=CC(=C(C=C1)F)[N+](=O)[O-]
CAS Min % 97.5
CAS Max % 100.0
Melting Point 46°C to 50°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 183.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4'-Fluoro-3'-nitroacetophenone
Show More Show Less
compliance-icons
Product Identifier
  • 4'-Fluoro-3'-nitroacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'