missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methoxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number p-7036476
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
425378-85-4
C13H18BFO3
252.09
MFCD11227055
WAEWRICTVAIONI-UHFFFAOYSA-N
2-4-fluoro-3-methoxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-fluoro-3-methoxyphenylboronic acid pinacol ester, 2-4-fluoro-3-methoxyphenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 4-fluoro-3-methoxybenzeneboronic acid pinacol ester, 2-4-fluoro-3-methoxy-phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4-fluoro-3-methoxyphenyl-4,4,5,5-tetramethyl 1,3,2 dioxaborolane
22285305
2-(4-fluoro-3-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
COC1=C(F)C=CC(=C1)B1OC(C)(C)C(C)(C)O1
This item is not returnable. View return policy
425378-85-4
C13H18BFO3
252.09
MFCD11227055
WAEWRICTVAIONI-UHFFFAOYSA-N
2-4-fluoro-3-methoxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-fluoro-3-methoxyphenylboronic acid pinacol ester, 2-4-fluoro-3-methoxyphenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 4-fluoro-3-methoxybenzeneboronic acid pinacol ester, 2-4-fluoro-3-methoxy-phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4-fluoro-3-methoxyphenyl-4,4,5,5-tetramethyl 1,3,2 dioxaborolane
22285305
2-(4-fluoro-3-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
COC1=C(F)C=CC(=C1)B1OC(C)(C)C(C)(C)O1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 69°C to 71°C
Quantity 250 mg
Formula Weight 252.09
Percent Purity 96%
Chemical Name or Material 4-Fluoro-3-methoxybenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'