missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-3-[methoxy(methyl)carbamoyl]benzeneboronic acid, 98%, Alfa Aesar™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Melting Point | 160°C to 163°C |
Quantity | 250 mg |
Formula Weight | 227 |
Percent Purity | 98% |
Chemical Name or Material | 4-Fluoro-3-[methoxy(methyl)carbamoyl]benzeneboronic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'