missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-hydroxybenzoic acid, 97%, Thermo Scientific™

Catalog Number p-4323351
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
51446-31-2
C7H4FO3
155.11
MFCD00269641
QATKOZUHTGAWMG-UHFFFAOYSA-M
benzoic acid, 4-fluoro-3-hydroxy, 4-fluoro-3-hydroxybenzoicacid, 4-fluoro-3-hydroxy-benzoic acid, 3-hydroxy-4-fluorobenzoic acid, pubchem3494, ksc497o4f, 3-hydroxy-4-fluoro-benzoic acid, 4-fluoro-3-hydroxy benzoic acid, timtec-bb sbb008505, buttpark 34\01-10
593635
4-fluoro-3-hydroxybenzoic acid
OC1=CC(=CC=C1F)C([O-])=O
This item is not returnable. View return policy
51446-31-2
C7H4FO3
155.11
MFCD00269641
QATKOZUHTGAWMG-UHFFFAOYSA-M
benzoic acid, 4-fluoro-3-hydroxy, 4-fluoro-3-hydroxybenzoicacid, 4-fluoro-3-hydroxy-benzoic acid, 3-hydroxy-4-fluorobenzoic acid, pubchem3494, ksc497o4f, 3-hydroxy-4-fluoro-benzoic acid, 4-fluoro-3-hydroxy benzoic acid, timtec-bb sbb008505, buttpark 34\01-10
593635
4-fluoro-3-hydroxybenzoic acid
OC1=CC(=CC=C1F)C([O-])=O
CAS Min % 96.0
CAS Max % 100.0
Melting Point 215.0°C to 217.0°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 156.11
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 4-Fluoro-3-hydroxybenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluoro-3-hydroxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'