missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-(trifluoromethyl)phenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-4228696
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
182344-16-7
C7H5BF4O2
207.92
MFCD08062379
SWUPLEAGZOKLNX-UHFFFAOYSA-N
4-fluoro-2-trifluoromethyl phenyl boronic acid, 4-fluoro-2-trifluoromethyl benzeneboronic acid, 4-fluoro-2-trifluoromethyl phenylboronic acid, 2-borono-5-fluorobenzotrifluoride, boronic acid, 4-fluoro-2-trifluoromethyl phenyl, akos brn-1027, acmc-1byq0, 4-fluoro-2-trifluoromethyl-phenylboronic acid, ksc495e5n
12109469
[4-fluoro-2-(trifluoromethyl)phenyl]boronic acid
OB(O)C1=C(C=C(F)C=C1)C(F)(F)F
This item is not returnable. View return policy
182344-16-7
C7H5BF4O2
207.92
MFCD08062379
SWUPLEAGZOKLNX-UHFFFAOYSA-N
4-fluoro-2-trifluoromethyl phenyl boronic acid, 4-fluoro-2-trifluoromethyl benzeneboronic acid, 4-fluoro-2-trifluoromethyl phenylboronic acid, 2-borono-5-fluorobenzotrifluoride, boronic acid, 4-fluoro-2-trifluoromethyl phenyl, akos brn-1027, acmc-1byq0, 4-fluoro-2-trifluoromethyl-phenylboronic acid, ksc495e5n
12109469
[4-fluoro-2-(trifluoromethyl)phenyl]boronic acid
OB(O)C1=C(C=C(F)C=C1)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 184.0°C to 186.0°C
Color Brown to Gray
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 207.92
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)phenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluoro-2-(trifluoromethyl)phenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'