missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-(trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific™

Catalog Number 10492884
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
189807-21-4
C8H3ClF4O
226.55
MFCD00061158
OEYHURRIOWWRMD-UHFFFAOYSA-N
4-fluoro-2-trifluoromethyl benzoyl chloride, 4-fluoro-2-trifluoromethyl benzoylchloride, 2-trifluoromethyl-4-fluorobenzoyl chloride, 2-chlorocarbonyl-5-fluorobenzotrifluoride, benzoyl chloride, 4-fluoro-2-trifluoromethyl, alpha,alpha,alpha,4-tetrafluoro-o-toluoyl chloride, acmc-1bwn8, ksc540e2h, attercop-chm at107069, 2-trifluoromethyl-4-fluorobenzoylchlorid
605680
4-fluoro-2-(trifluoromethyl)benzoyl chloride
FC1=CC=C(C(Cl)=O)C(=C1)C(F)(F)F
This item is not returnable. View return policy
189807-21-4
C8H3ClF4O
226.55
MFCD00061158
OEYHURRIOWWRMD-UHFFFAOYSA-N
4-fluoro-2-trifluoromethyl benzoyl chloride, 4-fluoro-2-trifluoromethyl benzoylchloride, 2-trifluoromethyl-4-fluorobenzoyl chloride, 2-chlorocarbonyl-5-fluorobenzotrifluoride, benzoyl chloride, 4-fluoro-2-trifluoromethyl, alpha,alpha,alpha,4-tetrafluoro-o-toluoyl chloride, acmc-1bwn8, ksc540e2h, attercop-chm at107069, 2-trifluoromethyl-4-fluorobenzoylchlorid
605680
4-fluoro-2-(trifluoromethyl)benzoyl chloride
FC1=CC=C(C(Cl)=O)C(=C1)C(F)(F)F
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.4950g/mL
Boiling Point 157.0°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CF3C6H3(F)COCl
Quantity 5 g
Specific Gravity 1.495
Formula Weight 226.56
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)benzoyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluoro-2-(trifluoromethyl)benzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'