missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-(trifluoromethyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number 15438076
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1416723-10-8
C13H15BF4O2
290.065
MPWDTDMRMPOPDS-UHFFFAOYSA-N
4-fluoro-2-trifluoromethyl phenylboronic acid pinacol ester, 2-4-fluoro-2-trifluoromethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
100913434
2-[4-fluoro-2-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)F)C(F)(F)F
This item is not returnable. View return policy
1416723-10-8
C13H15BF4O2
290.065
MPWDTDMRMPOPDS-UHFFFAOYSA-N
4-fluoro-2-trifluoromethyl phenylboronic acid pinacol ester, 2-4-fluoro-2-trifluoromethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
100913434
2-[4-fluoro-2-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)F)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 290.06
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'