missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Fluoro-2-(trifluoromethoxy)phenylacetonitrile, 97%

Brand:  Alfa Aesar™ H34369.03

Code : NEW

Additional Details : Weight : 0.00100kg Transport : UN number : 3276 Chem class : 6.1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15448905

  • £165.00 / 1g


Estimated Shipment date
from Supplier 06-07-2018
Add to basket

Specifications

Chemical Identifiers

4-Fluoro-2-(trifluoromethoxy)phenylacetonitrile
219.14
MFCD19687176
UN3276

Specifications

4-Fluoro-2-(trifluoromethoxy)phenylacetonitrile
1323966-14-8
97%
MFCD19687176
1g
Liquid
219.14
UN3276
Videos
SDS