missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-nitroaniline, 99%, Thermo Scientific Chemicals

Catalog Number p-135381
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
364-78-3
C6H5FN2O2
156.12
MFCD00007830
PUGDHSSOXPHLPT-UHFFFAOYSA-N
2-nitro-4-fluoroaniline, 4-fluoro-2-nitrophenylamine, 4-fluoro-2-nitrobenzeneamine, 4-fluoro-2-nitrobenzenamine, benzenamine, 4-fluoro-2-nitro, 2-amino-5-fluoronitrobenzene, 4-fluoro-2-nitro-aniline, 4-fluoro-2-nitro-phenylamine, 4-fluor-2-nitroanilin, pubchem3421
67769
4-fluoro-2-nitroaniline
NC1=CC=C(F)C=C1[N+]([O-])=O
This item is not returnable. View return policy
364-78-3
C6H5FN2O2
156.12
MFCD00007830
PUGDHSSOXPHLPT-UHFFFAOYSA-N
2-nitro-4-fluoroaniline, 4-fluoro-2-nitrophenylamine, 4-fluoro-2-nitrobenzeneamine, 4-fluoro-2-nitrobenzenamine, benzenamine, 4-fluoro-2-nitro, 2-amino-5-fluoronitrobenzene, 4-fluoro-2-nitro-aniline, 4-fluoro-2-nitro-phenylamine, 4-fluor-2-nitroanilin, pubchem3421
67769
4-fluoro-2-nitroaniline
NC1=CC=C(F)C=C1[N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 92°C to 95°C
Color Orange to Brown
Flash Point 89°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula FC6H3(NO2)NH2
Quantity 10 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in polar organic solvents,insoluble in non-polar organic solvents,approx. 12% soluble in alcohol (15°C)
Formula Weight 156.12
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-2-nitroaniline, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'