missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluoro-2'-nitroacetanilide, 99+%, Thermo Scientific™

Catalog Number p-135354
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
448-39-5
C8H7FN2O3
198.16
MFCD00031517
UZBZEUCQENVPQB-UHFFFAOYSA-N
4'-fluoro-2'-nitroacetanilide, n-4-fluoro-2-nitrophenyl acetamide, 4-fluoro-2-nitroacetanilide, maybridge1_003501, 4/'-fluoro-2/'-nitroacetanilide, labotest-bb lt00455473, labotest-bb lt03332301, acetamide,n-4-fluoro-2-nitrophenyl, n-4-fluoro-2-nitro-phenyl-acetamide, n-4-fluoro-2-nitrophenyl acetamide #
136289
N-(4-fluoro-2-nitrophenyl)acetamide
CC(=O)NC1=C(C=C(C=C1)F)[N+](=O)[O-]
This item is not returnable. View return policy
448-39-5
C8H7FN2O3
198.16
MFCD00031517
UZBZEUCQENVPQB-UHFFFAOYSA-N
4'-fluoro-2'-nitroacetanilide, n-4-fluoro-2-nitrophenyl acetamide, 4-fluoro-2-nitroacetanilide, maybridge1_003501, 4/'-fluoro-2/'-nitroacetanilide, labotest-bb lt00455473, labotest-bb lt03332301, acetamide,n-4-fluoro-2-nitrophenyl, n-4-fluoro-2-nitro-phenyl-acetamide, n-4-fluoro-2-nitrophenyl acetamide #
136289
N-(4-fluoro-2-nitrophenyl)acetamide
CC(=O)NC1=C(C=C(C=C1)F)[N+](=O)[O-]
CAS Min % 99.0
CAS Max % 100.0
Melting Point 70°C to 73°C
Color Ochre to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula FC6H3NO2NHC(O)CH3
Quantity 5 g
Solubility Information Solubility in water: soluble in hot water. Other solubilities: insoluble in non-polar solvents,soluble in polar,solvents
Formula Weight 198.16
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 4'-Fluoro-2'-nitroacetanilide, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 4'-Fluoro-2'-nitroacetanilide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'