missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-nitro-N,N-di-n-propylaniline, 97%, Thermo Scientific™

Catalog Number 15460399
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
(4-Fluoro-2-nitrophenyl)di-n-propylamine
This item is not returnable. View return policy
(4-Fluoro-2-nitrophenyl)di-n-propylamine

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN2811
Formula Weight 240.27
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Fluoro-2-nitro-N,N-di-n-propylaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'