missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Fluoro-2-n-propoxybenzeneboronic acid, 97%

Brand:  Alfa Aesar™ H55269.03

Code : NEW

Additional Details : CAS Number : 480438-60-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15451468

  • £330.93 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

4-Fluoro-2-n-propoxybenzeneboronic acid
480438-60-6
198
MFCD06411288

Specifications

4-Fluoro-2-n-propoxybenzeneboronic acid
480438-60-6
97%
MFCD06411288
1g
Solid
198
Videos
SDS