missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylpyridine, 96%, Thermo Scientific Chemicals

Catalog Number 15461409
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
766-16-5
C6H6FN
111.119
MFCD04114145
LEJYGRVERQVBLB-UHFFFAOYSA-N
4-fluoro-2-picoline, 4-fluoro-2-methyl-pyridine, pubchem21038, pyridine,4-fluoro-2-methyl, 4-fluoranyl-2-methyl-pyridine
2762819
4-fluoro-2-methylpyridine
CC1=NC=CC(=C1)F
This item is not returnable. View return policy
766-16-5
C6H6FN
111.119
MFCD04114145
LEJYGRVERQVBLB-UHFFFAOYSA-N
4-fluoro-2-picoline, 4-fluoro-2-methyl-pyridine, pubchem21038, pyridine,4-fluoro-2-methyl, 4-fluoranyl-2-methyl-pyridine
2762819
4-fluoro-2-methylpyridine
CC1=NC=CC(=C1)F

It is used in the correlations with and limitations of the α scale of solvent hydrogen bond donor acidities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the correlations with and limitations of the α scale of solvent hydrogen bond donor acidities.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store away from air, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Boiling Point 128°C to 132°C
Refractive Index 1.4715
Quantity 250 mg
Beilstein 107087
Sensitivity Air sensitive
Solubility Information Slightly soluble in water.
Formula Weight 111.12
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 4-Fluoro-2-methylpyridine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'