missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-2-methylpyridine, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
It is used in the correlations with and limitations of the α scale of solvent hydrogen bond donor acidities.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the correlations with and limitations of the α scale of solvent hydrogen bond donor acidities.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Store away from air, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store
It is used in the correlations with and limitations of the α scale of solvent hydrogen bond donor acidities.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Store away from air, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store
Specifications
Color | Colorless to Yellow |
Boiling Point | 128°C to 132°C |
Refractive Index | 1.4715 |
Quantity | 5 g |
Beilstein | 107087 |
Sensitivity | Air sensitive |
Solubility Information | Slightly soluble in water. |
Formula Weight | 111.12 |
Percent Purity | 96% |
Physical Form | Liquid |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'