missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylphenol, 98%, Thermo Scientific™

Catalog Number p-134705
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
452-72-2
C7H7FO
126.13
MFCD00075088
GKQDDKKGDIVDAG-UHFFFAOYSA-N
2-methyl-4-fluorophenol, 4-fluoro-o-cresol, 4-fluoro-2-methyl-phenol, 5-fluoro-2-hydroxytoluene, phenol, 4-fluoro-2-methyl, pubchem1500, acmc-1csjj, 4-fuoro-2-methyl-phenol, 2-methyl-4-fluoro phenol, 4-fluoro-2-methyl phenol
136295
4-fluoro-2-methylphenol
CC1=C(C=CC(=C1)F)O
This item is not returnable. View return policy
452-72-2
C7H7FO
126.13
MFCD00075088
GKQDDKKGDIVDAG-UHFFFAOYSA-N
2-methyl-4-fluorophenol, 4-fluoro-o-cresol, 4-fluoro-2-methyl-phenol, 5-fluoro-2-hydroxytoluene, phenol, 4-fluoro-2-methyl, pubchem1500, acmc-1csjj, 4-fuoro-2-methyl-phenol, 2-methyl-4-fluoro phenol, 4-fluoro-2-methyl phenol
136295
4-fluoro-2-methylphenol
CC1=C(C=CC(=C1)F)O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 37°C to 38°C
Color Brown to White
Boiling Point 87°C (14.0 mmHg)
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H3(CH3)OH
Refractive Index 1.5155 to 1.5175
Quantity 1 g
Formula Weight 126.13
Percent Purity 98%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 4-Fluoro-2-methylphenol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluoro-2-methylphenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'