missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylbenzoic acid, 99%, Thermo Scientific™

Catalog Number p-3771009
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
321-21-1
C8H7FO2
154.14
MFCD04115880
KDXOONIQRUZGSY-UHFFFAOYSA-N
4-fluoro-o-toluic acid, 2-methyl-4-fluorobenzoic acid, 4-fluoro-2-methylbenzoicacid, 4-fluoro-2-methyl-benzoic acid, benzoic acid, 4-fluoro-2-methyl, pubchem1327, 2-carboxy-5-fluorotoluene, acmc-209hr6, 4-fluoro-2-methyl benzoicacid, rarechem al be 0506
3860639
4-fluoro-2-methylbenzoic acid
CC1=C(C=CC(=C1)F)C(=O)O
This item is not returnable. View return policy
321-21-1
C8H7FO2
154.14
MFCD04115880
KDXOONIQRUZGSY-UHFFFAOYSA-N
4-fluoro-o-toluic acid, 2-methyl-4-fluorobenzoic acid, 4-fluoro-2-methylbenzoicacid, 4-fluoro-2-methyl-benzoic acid, benzoic acid, 4-fluoro-2-methyl, pubchem1327, 2-carboxy-5-fluorotoluene, acmc-209hr6, 4-fluoro-2-methyl benzoicacid, rarechem al be 0506
3860639
4-fluoro-2-methylbenzoic acid
CC1=C(C=CC(=C1)F)C(=O)O
Melting Point 168.0°C to 172.0°C
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula FC6H3(CH3)COOH
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 154.14
Percent Purity 99%
Chemical Name or Material 4-Fluoro-2-methylbenzoic acid
compliance-icons
Product Identifier
  • 4-Fluoro-2-methylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'