missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylbenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number 15457056
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
815631-56-2
C13H18BFO2
236.09
MFCD05863911
HHWOQSZBVGPYMH-UHFFFAOYSA-N
2-4-fluoro-2-methylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-fluoro-2-methylphenylboronic acid pinacol ester, 4-fluoro-2-methylphenylboronic acid, pinacol ester, 2-4-fluoro-2-methylphenyl 4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-fluoro-2-methylphenylboronic acid,pinacol ester, 4-fluoro-2-methylbenzeneboronic acid pinacol ester, 2-4-fluoranyl-2-methyl-phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4-fluoro-2-methyl-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 1,3,2-dioxaborolane, 2-4-fluoro-2-methylphenyl-4,4,5,5-tetramethyl
2759037
2-(4-fluoro-2-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CC1=CC(F)=CC=C1B1OC(C)(C)C(C)(C)O1
This item is not returnable. View return policy
815631-56-2
C13H18BFO2
236.09
MFCD05863911
HHWOQSZBVGPYMH-UHFFFAOYSA-N
2-4-fluoro-2-methylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-fluoro-2-methylphenylboronic acid pinacol ester, 4-fluoro-2-methylphenylboronic acid, pinacol ester, 2-4-fluoro-2-methylphenyl 4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-fluoro-2-methylphenylboronic acid,pinacol ester, 4-fluoro-2-methylbenzeneboronic acid pinacol ester, 2-4-fluoranyl-2-methyl-phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4-fluoro-2-methyl-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 1,3,2-dioxaborolane, 2-4-fluoro-2-methylphenyl-4,4,5,5-tetramethyl
2759037
2-(4-fluoro-2-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CC1=CC(F)=CC=C1B1OC(C)(C)C(C)(C)O1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 236.09
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 4-Fluoro-2-methylbenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'