missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methoxyphenylboronic acid, 97%, ACROS Organics™

Brand:  Acros Organics 434080050

Code : 29

Additional Details : CAS Number : 179899-07-1 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10696840

  • £107.10 / 5g


Estimated Shipment
09-02-2018
Add to basket


Specifications

Chemical Identifiers

4-Fluoro-2-methoxyphenylboronic acid
96.0
≥96.0%
C7H8BFO3
MFCD03788423
179899-07-1
100.0
97%
169.95

Specifications

4-Fluoro-2-methoxyphenylboronic acid
5g
96.0
100.0
Crystalline Powder
97%
169.95
White
179899-07-1
Authentic
Glass bottle
≥96.0%
C7H8BFO3
MFCD03788423
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Fluoro-2-methoxyphenylboronic acid, 97%

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ A§ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS