missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methoxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number 15477366
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
624741-83-9
C13H18BFO3
252.092
IYTUTWZXSBERDH-UHFFFAOYSA-N
2-4-fluoro-2-methoxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-fluoro-2-methoxyphenylboronic acid pinacol ester, 2-4-fluoro-2-methoxyphenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane
22240091
2-(4-fluoro-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)F)OC
This item is not returnable. View return policy
624741-83-9
C13H18BFO3
252.092
IYTUTWZXSBERDH-UHFFFAOYSA-N
2-4-fluoro-2-methoxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-fluoro-2-methoxyphenylboronic acid pinacol ester, 2-4-fluoro-2-methoxyphenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane
22240091
2-(4-fluoro-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)F)OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 252.09
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 4-Fluoro-2-methoxybenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'