missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Fluoro-2-isopropoxybenzeneboronic acid, 97%

Brand:  Alfa Aesar™ H55338.03

Code : NEW

Additional Details : CAS Number : 480438-59-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15481558

  • £264.70 / 1g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Fluoro-2-isopropoxybenzeneboronic acid
480438-59-3
198
MFCD06411287

Specifications

4-Fluoro-2-isopropoxybenzeneboronic acid
480438-59-3
97%
MFCD06411287
1g
Solid
198
Videos
SDS