missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-hydroxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number 15467206
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1038828-32-8
C12H16BFO3
238.07
MFCD16994248
PSQUBOBJDBVJIN-UHFFFAOYSA-N
5-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-fluoro-2-hydroxybenzeneboronic acid pinacol ester, 4-fluoro-2-hydroxyphenylboronic acid pinacol ester, phenol, 5-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
72219414
5-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
CC1(C)OB(OC1(C)C)C1=C(O)C=C(F)C=C1
This item is not returnable. View return policy
1038828-32-8
C12H16BFO3
238.07
MFCD16994248
PSQUBOBJDBVJIN-UHFFFAOYSA-N
5-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-fluoro-2-hydroxybenzeneboronic acid pinacol ester, 4-fluoro-2-hydroxyphenylboronic acid pinacol ester, phenol, 5-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
72219414
5-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
CC1(C)OB(OC1(C)C)C1=C(O)C=C(F)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 238.06
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 4-Fluoro-2-hydroxybenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'