missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Fluoro-2-formylbenzeneboronic acid pinacol ester, 96%

Brand:  Alfa Aesar™ H62104.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15407036

  • £283.80 / 1g


Estimated Shipment date
from Supplier 04-05-2018
Add to basket

Specifications

Chemical Identifiers

4-Fluoro-2-formylbenzeneboronic acid pinacol ester
250.07
4-Fluoro-2-formylphenylboronic acid pinacol ester

Specifications

4-Fluoro-2-formylbenzeneboronic acid pinacol ester
Solid
250.07
1g
96%
4-Fluoro-2-formylphenylboronic acid pinacol ester
Videos
SDS