missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-formylbenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number 15417036
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
943310-52-9
C13H16BFO3
250.076
STGVEURXAACTES-UHFFFAOYSA-N
5-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 4-fluoro-2-formylbenzeneboronicacidpinacolester, 4-fluoro-2-formylbenzeneboronic acid pinacol ester, 5-fluoro-2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzaldehyde
59420279
5-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)F)C=O
This item is not returnable. View return policy
943310-52-9
C13H16BFO3
250.076
STGVEURXAACTES-UHFFFAOYSA-N
5-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 4-fluoro-2-formylbenzeneboronicacidpinacolester, 4-fluoro-2-formylbenzeneboronic acid pinacol ester, 5-fluoro-2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzaldehyde
59420279
5-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)F)C=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 250.07
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 4-Fluoro-2-formylbenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'