missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-(1H-pyrazol-3-yl)phenol, 97%, Thermo Scientific™

Catalog Number p-4229219
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
288401-64-9
C9H7FN2O
178.17
DIVXDXZKHCYBPY-FPLPWBNLSA-N
4-fluoro-2-1h-pyrazol-3-yl phenol, 3-5-fluoro-2-hydroxyphenyl pyrazole, pyrazole, 31, 6-3z-1,2-dihydropyrazol-3-ylidene-4-fluorocyclohexa-2,4-dien-1-one
16217010
(6Z)-6-(1,2-dihydropyrazol-3-ylidene)-4-fluorocyclohexa-2,4-dien-1-one
C1=CC(=O)C(=C2C=CNN2)C=C1F
This item is not returnable. View return policy
288401-64-9
C9H7FN2O
178.17
DIVXDXZKHCYBPY-FPLPWBNLSA-N
4-fluoro-2-1h-pyrazol-3-yl phenol, 3-5-fluoro-2-hydroxyphenyl pyrazole, pyrazole, 31, 6-3z-1,2-dihydropyrazol-3-ylidene-4-fluorocyclohexa-2,4-dien-1-one
16217010
(6Z)-6-(1,2-dihydropyrazol-3-ylidene)-4-fluorocyclohexa-2,4-dien-1-one
C1=CC(=O)C(=C2C=CNN2)C=C1F
CAS Min % 96.0
CAS Max % 100.0
Melting Point 132.9°C to 134.1°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 178.17
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 4-Fluoro-2-(1H-pyrazol-3-yl)phenol
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluoro-2-(1H-pyrazol-3-yl)phenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'