missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-1,3-phenylenediamine, 95%, Thermo Scientific Chemicals

Catalog Number p-7053795
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
6264-67-1
C6H7FN2
126.13
MFCD00077494
QNDFYLBDUWCFJO-UHFFFAOYSA-N
80435
4-fluorobenzene-1,3-diamine
NC1=CC=C(F)C(N)=C1
This item is not returnable. View return policy
6264-67-1
C6H7FN2
126.13
MFCD00077494
QNDFYLBDUWCFJO-UHFFFAOYSA-N
80435
4-fluorobenzene-1,3-diamine
NC1=CC=C(F)C(N)=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 34°C to 37°C
Color Black
Boiling Point 152°C to 153°C
Flash Point 105°C (221°F)
Quantity 1 g
Synonym 2,4-diaminofluorobenzene, 4-fluoro-1,3-benzenediamine, 4-fluoro-1,3-phenylenediamine, 1,3-diamino-4-fluorobenzene, 1,3-benzenediamine, 4-fluoro, 4-fluoro-1,3-diaminobenzene, 1,3-benzenediamine,4-fluoro, pubchem2538, acmc-209n6n, ksc615i4l
Formula Weight 126.13
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 4-Fluoro-1,3-phenylenediamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'