missing translation for 'onlineSavingsMsg'
Learn More

4-Ethynylbenzonitrile, 97%, Thermo Scientific Chemicals

Catalog Number 15477748
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3032-92-6
C9H5N
127.146
MFCD04974058
LAGNMUUUMQJXBF-UHFFFAOYSA-N
4-ethynyl-benzonitrile, 4-cyanophenylacetylene, benzonitrile, 4-ethynyl, p-cyanophenylacetylene, 4-ethynyl-bezonitrile, benzonitrile,4-ethynyl, 4-ethynylbenzenecarbonitrile, 4-ethynylbenzonitrile, ksc564s8b, benzonitrile, 4-ethynyl-9ci
4547245
4-ethynylbenzonitrile
C#CC1=CC=C(C=C1)C#N
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3032-92-6
C9H5N
127.146
MFCD04974058
LAGNMUUUMQJXBF-UHFFFAOYSA-N
4-ethynyl-benzonitrile, 4-cyanophenylacetylene, benzonitrile, 4-ethynyl, p-cyanophenylacetylene, 4-ethynyl-bezonitrile, benzonitrile,4-ethynyl, 4-ethynylbenzenecarbonitrile, 4-ethynylbenzonitrile, ksc564s8b, benzonitrile, 4-ethynyl-9ci
4547245
4-ethynylbenzonitrile
C#CC1=CC=C(C=C1)C#N

4-Ethynylbenzonitrile is used as a synthetic fragment and as a test compound for cross-coupling reactions. It is used in the study of hydrogen bond formation in multifunctional molecules due to the presence of four hydrogen bonding sites. It is also involved in the preparation of 4-[(trimethylsilyl)ethynyl]benzonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Ethynylbenzonitrile is used as a synthetic fragment and as a test compound for cross-coupling reactions. It is used in the study of hydrogen bond formation in multifunctional molecules due to the presence of four hydrogen bonding sites. It is also involved in the preparation of 4-[(trimethylsilyl)ethynyl]benzonitrile.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents, acids and organic materials.
TRUSTED_SUSTAINABILITY
Melting Point 156°C to 160°C
Color Yellow
Quantity 1 g
UN Number UN3439
Solubility Information Slightly soluble in water.
Formula Weight 127.14
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Ethynylbenzonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'