missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylthiophenol, 97%, Thermo Scientific Chemicals

Catalog Number p-7027949
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
4946-13-8
C8H9S
137.22
MFCD00040229
WWQQPHUHTAZWDH-UHFFFAOYSA-M
4-ethylthiophenol, 4-ethyl thiophenol, 4-ethylbenzene-1-thiol, benzenethiol, 4-ethyl, p-ethylphenylmercaptan, p-ethylthiophenol, pubchem6813, p-ethylphenyl mercaptan, acmc-209kg5
2758914
4-ethylbenzenethiol
CCC1=CC=C([S-])C=C1
This item is not returnable. View return policy
4946-13-8
C8H9S
137.22
MFCD00040229
WWQQPHUHTAZWDH-UHFFFAOYSA-M
4-ethylthiophenol, 4-ethyl thiophenol, 4-ethylbenzene-1-thiol, benzenethiol, 4-ethyl, p-ethylphenylmercaptan, p-ethylthiophenol, pubchem6813, p-ethylphenyl mercaptan, acmc-209kg5
2758914
4-ethylbenzenethiol
CCC1=CC=C([S-])C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 119°C to 120°C (30 mmHg)
Odor Stench
Refractive Index 1.5645
Quantity 1 g
UN Number UN2810
Beilstein 1905734
Sensitivity Air Sensitive
Formula Weight 138.23
Percent Purity 97%
Chemical Name or Material 4-Ethylthiophenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'