missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylpyridine 98%, ACROS Organics™

Brand:  Acros Organics 150571000

Code : 29

Additional Details : CAS Number : 536-75-4 Weight : 0.15000kg Transport : UN number : 1992 Chem class : 3 Pack group : III


Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10521591

  • £40.60 / 100g


Estimated Shipment
13-07-2018
Add to basket


Specifications

Chemical Identifiers

4-Ethylpyridine
536-75-4
100.0
98%
(C5H3N)CH2CH3
MFCD00006450
98%
97.5
≥97.5%
C7H9N
107.15

Specifications

4-Ethylpyridine
100g
536-75-4
Authentic
0.942g/mL
1.5009 to 1.5029 (20°C, 589nm)
Liquid
98%
(C5H3N)CH2CH3
MFCD00006450
15, 3901
Clear colorless to yellow
98%
97.5
100.0
Glass bottle
0.5% max. (K.F.)
≥97.5%
C7H9N
107.15
20, 243
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Ethylpyridine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS