missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylcarbamoyl-3-fluorobenzeneboronic acid, 98%, Thermo Scientific™

Catalog Number p-7035555
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
874289-12-0
C9H11BFNO3
211.00
MFCD08436070
PCACUISZWVJQCI-UHFFFAOYSA-N
4-ethylcarbamoyl-3-fluorophenyl boronic acid, n-ethyl 4-borono-2-fluorobenzamide, 4-ethylcarbamoyl-3-fluorobenzeneboronic acid, 4-ethylcarbamoyl-3-fluorophenylboronic acid, boronic acid,b-4-ethylamino carbonyl-3-fluorophenyl, acmc-209qm5, n-ethyl-4-borono-2-fluorobenzamide, 4-ethylcarbamoyl-3-fluorophenyl boronicacid, 4-ethylcarbamoyl-3-fluoranyl-phenyl boronic acid
44717595
[4-(ethylcarbamoyl)-3-fluorophenyl]boronic acid
CCNC(=O)C1=C(F)C=C(C=C1)B(O)O
This item is not returnable. View return policy
874289-12-0
C9H11BFNO3
211.00
MFCD08436070
PCACUISZWVJQCI-UHFFFAOYSA-N
4-ethylcarbamoyl-3-fluorophenyl boronic acid, n-ethyl 4-borono-2-fluorobenzamide, 4-ethylcarbamoyl-3-fluorobenzeneboronic acid, 4-ethylcarbamoyl-3-fluorophenylboronic acid, boronic acid,b-4-ethylamino carbonyl-3-fluorophenyl, acmc-209qm5, n-ethyl-4-borono-2-fluorobenzamide, 4-ethylcarbamoyl-3-fluorophenyl boronicacid, 4-ethylcarbamoyl-3-fluoranyl-phenyl boronic acid
44717595
[4-(ethylcarbamoyl)-3-fluorophenyl]boronic acid
CCNC(=O)C1=C(F)C=C(C=C1)B(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 157°C to 160°C
Quantity 250 mg
Formula Weight 211
Percent Purity 98%
Chemical Name or Material 4-Ethylcarbamoyl-3-fluorobenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'