missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylaniline, 99+%, Thermo Scientific Chemicals

Catalog Number 10667642
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
589-16-2
C8H11N
121.18
MFCD00007921
HRXZRAXKKNUKRF-UHFFFAOYSA-N
p-ethylaniline, benzenamine, 4-ethyl, 4-ethylphenylamine, aniline, 4-ethyl, p-ethylaminobenzene, 1-amino-4-ethylbenzene, 4-aminoethylbenzene, aniline, p-ethyl, unii-mhx72w1zqv, ccris 5060
11504
4-ethylaniline
CCC1=CC=C(N)C=C1
This item is not returnable. View return policy
589-16-2
C8H11N
121.18
MFCD00007921
HRXZRAXKKNUKRF-UHFFFAOYSA-N
p-ethylaniline, benzenamine, 4-ethyl, 4-ethylphenylamine, aniline, 4-ethyl, p-ethylaminobenzene, 1-amino-4-ethylbenzene, 4-aminoethylbenzene, aniline, p-ethyl, unii-mhx72w1zqv, ccris 5060
11504
4-ethylaniline
CCC1=CC=C(N)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point -5°C
Color Brown-Red to Yellow
Density 0.9700g/mL
Boiling Point 216°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula C2H5C6H4NH2
Refractive Index 1.5532 to 1.5552
Quantity 100 g
Beilstein 12, 1090
Specific Gravity 0.97
Solubility Information Solubility in water: immiscible
Formula Weight 121.18
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 4-Ethylaniline, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Ethylaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'