4-ethyl hydrogen 1-methyl-1H-pyrazole-4,5-dicarboxylate, Maybridge


Overview
Brand: Maybridge

Manufacturer Part Number: CC76801DA

UNSPSC: 12350000

Code: 31

Additional Details:
Additional Details: CAS Number: 81303-65-3 Weight: 0.05100kg


Packaging Amber Glass Bottle
Quantity 1g

Product Code. 10643625

 N/A
N/A

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description and Specification

Chemical Identifiers

Chemical Name or Material 4-ethyl hydrogen 1-methyl-1H-pyrazole-4, 5-dicarboxylate
Assay 97%
Molecular Formula C8H10N2O4
Formula Weight 198.18

Specification

Formula Weight 198.18
Quantity 1g
Assay 97%
CAS 81303-65-3
Packaging Amber Glass Bottle
Chemical Name or Material 4-ethyl hydrogen 1-methyl-1H-pyrazole-4, 5-dicarboxylate
Molecular Formula C8H10N2O4
Documentation

Technical documents