missing translation for 'onlineSavingsMsg'
Learn More

4-Ethyl-2-methoxyphenol, 98%, Thermo Scientific Chemicals

Catalog Number p-130757
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2785-89-9
C9H12O2
152.19
MFCD00038714
CHWNEIVBYREQRF-UHFFFAOYSA-N
4-ethylguaiacol, p-ethylguaiacol, 2-methoxy-4-ethylphenol, phenol, 4-ethyl-2-methoxy, homocresol, guaiacyl ethane, 4-ethyl-2-methoxy-phenol, 4-hydroxy-3-methoxy ethylbenzene, unii-c9nfd83bj5, guaiacol, 4-ethyl
62465
4-ethyl-2-methoxyphenol
CCC1=CC=C(O)C(OC)=C1
This item is not returnable. View return policy
2785-89-9
C9H12O2
152.19
MFCD00038714
CHWNEIVBYREQRF-UHFFFAOYSA-N
4-ethylguaiacol, p-ethylguaiacol, 2-methoxy-4-ethylphenol, phenol, 4-ethyl-2-methoxy, homocresol, guaiacyl ethane, 4-ethyl-2-methoxy-phenol, 4-hydroxy-3-methoxy ethylbenzene, unii-c9nfd83bj5, guaiacol, 4-ethyl
62465
4-ethyl-2-methoxyphenol
CCC1=CC=C(O)C(OC)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 15°C
Color Colorless to Yellow
Density 1.0580g/mL
Boiling Point 234°C to 236°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5250 to 1.53
Quantity 5 g
Specific Gravity 1.058
Solubility Information Solubility in water: insoluble
Formula Weight 152.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Ethyl-2-methoxyphenol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Ethyl-2-methoxyphenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'