missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Ethoxymethylene-2-phenyloxazolin-5-one, 98+%, Thermo Scientific Chemicals
19.40 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
25 g
Description
4-Ethoxymethylene-2-phenyloxazolin-5-one acts as sensitizing agent. It is also used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Ethoxymethylene-2-phenyloxazolin-5-one acts as sensitizing agent. It is also used as pharmaceutical intermediates.
Solubility
Soluble in methanol (50 mg/ml), and water (partly).
Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
4-Ethoxymethylene-2-phenyloxazolin-5-one acts as sensitizing agent. It is also used as pharmaceutical intermediates.
Solubility
Soluble in methanol (50 mg/ml), and water (partly).
Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
Specifications
Melting Point | 93°C to 97°C |
Quantity | 1 g |
Beilstein | 163055 |
Solubility Information | Soluble in methanol (50mg/ml),and water (partly). |
Formula Weight | 217.22 |
Percent Purity | ≥98% |
Chemical Name or Material | 4-Ethoxymethylene-2-phenyloxazolin-5-one |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'