missing translation for 'onlineSavingsMsg'
Learn More

4-(Ethoxycarbonyl)phenylzinc iodide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog Number p-7045937
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
1205-06-7
C10H9NO2S
207.247
MFCD00022058
MLOJHUCMCKBDLV-UHFFFAOYSA-N
4-ethoxycarbonylphenyl isothiocyanate, 4-ethoxycarbonylphenylisothiocyanate, benzoic acid, 4-isothiocyanato-, ethyl ester, 4-ethoxycarbonyl phenyl isothiocyanate, ethyl p-isothiocyanatobenzoate, 4-ethoxycarbonylphenyl-isothiocyanate, benzoic acid,4-isothiocyanato-, ethyl ester, acmc-1brcn, p-carbethoxyphenylisothiocyanate, #
71003
ethyl 4-isothiocyanatobenzoate
CCOC(=O)C1=CC=C(C=C1)N=C=S
This item is not returnable. View return policy
1205-06-7
C10H9NO2S
207.247
MFCD00022058
MLOJHUCMCKBDLV-UHFFFAOYSA-N
4-ethoxycarbonylphenyl isothiocyanate, 4-ethoxycarbonylphenylisothiocyanate, benzoic acid, 4-isothiocyanato-, ethyl ester, 4-ethoxycarbonyl phenyl isothiocyanate, ethyl p-isothiocyanatobenzoate, 4-ethoxycarbonylphenyl-isothiocyanate, benzoic acid,4-isothiocyanato-, ethyl ester, acmc-1brcn, p-carbethoxyphenylisothiocyanate, #
71003
ethyl 4-isothiocyanatobenzoate
CCOC(=O)C1=CC=C(C=C1)N=C=S

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 52°C to 56°C
Boiling Point 123°C to 127°C (1 mmHg)
Odor Irritating
Quantity 2 g
UN Number UN2923
Beilstein 2213400
Sensitivity Moisture sensitive
Formula Weight 207.25
Percent Purity 97%
Chemical Name or Material 4-(Ethoxycarbonyl)phenyl isothiocyanate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'