missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(Ethoxycarbonyl)phenyl isocyanate, 97%

Brand:  Alfa Aesar™ L10681.03

Code : NEW

Additional Details : CAS Number : 30806-83-8 Weight : 0.00100kg Transport : UN number : 2811 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 11322936

  • £17.17 / 1g


Estimated Shipment date
from Supplier 25-01-2018
Add to basket

Description

Description

Applications
Ethyl 4-isocyanatobenzoate was used in the preparation of cellulose carbamate and ester derivatives. It was also used in the preparation of ethyl 4-(3-(4-oxo-6-tridecyl-1,4-dihydropyrimidin-2-yl)ureido)benzoate2 and ethyl 4-(2-oxocyclohexanecarboxamido)benzoate.

Solubility
Hydrolyses in water.

Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from moisture, alocohol, amines, water.
Specifications

Chemical Identifiers

4-(Ethoxycarbonyl)phenyl isocyanate
C10H9NO3
MFCD00002027
882021
30806-83-8
191.19
UN2811
4-Carboethoxyphenyl isocyanate; Ethyl 4-isocyanatobenzoate

Specifications

4-(Ethoxycarbonyl)phenyl isocyanate
118° to 119°C (0.8mmHg)
1g
97%
191.19
UN2811
4-Carboethoxyphenyl isocyanate; Ethyl 4-isocyanatobenzoate
>110°C (230°F)
28° to 32°C
30806-83-8
C10H9NO3
MFCD00002027
882021
Videos
SDS